Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 11, 12 | 1462 | ||
NM_001040716.1 | NP_001035806.1 | 12, 13 | 1650 | |||
NM_022172.2 | NP_071504.2 | 10, 11 | 1417 | |||
XM_005274031.4 | XP_005274088.1 | 10, 11 | 1509 | |||
XM_005274032.4 | XP_005274089.1 | 10, 11 | 1391 | |||
XM_006718578.3 | XP_006718641.1 | 10, 11 | 1462 | |||
XM_011545086.2 | XP_011543388.1 | 10, 11 | 1778 | |||
XM_017017868.1 | XP_016873357.1 | 13, 14 | 1949 | |||
XM_017017869.1 | XP_016873358.1 | 13, 14 | 1820 | |||
XM_017017870.1 | XP_016873359.1 | 12, 13 | 1761 | |||
XM_017017871.1 | XP_016873360.1 | 12, 13 | 1632 | |||
XM_017017872.1 | XP_016873361.1 | 11, 12 | 2014 | |||
GenBank mRNA | BC011617.2 | - | 11, 12 | 1451 | 3946441 | |
S72370.1 | - | 10, 11 | 1450 | |||
U04641.1 | - | 10, 11 | 1406 | |||
U30891.1 | - | 11, 12 | 1420 |