Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000920.3 | NP_000911.2 | 18 | 2682 | ||
NM_001040716.1 | NP_001035806.1 | 19 | 2870 | |||
NM_022172.2 | NP_071504.2 | 17 | 2637 | |||
XM_005274031.4 | XP_005274088.1 | 17 | 2729 | |||
XM_005274032.4 | XP_005274089.1 | 17 | 2611 | |||
XM_006718578.3 | XP_006718641.1 | 17 | 2682 | |||
XM_006718579.3 | XP_006718642.1 | 9 | 1471 | |||
XM_011545086.2 | XP_011543388.1 | 17 | 2998 | |||
XM_011545087.2 | XP_011543389.1 | 9 | 1596 | |||
XM_017017868.1 | XP_016873357.1 | 20 | 3169 | |||
XM_017017869.1 | XP_016873358.1 | 20 | 3040 | |||
XM_017017870.1 | XP_016873359.1 | 19 | 2981 | |||
XM_017017871.1 | XP_016873360.1 | 19 | 2852 | |||
XM_017017872.1 | XP_016873361.1 | 18 | 3234 | |||
GenBank mRNA | BC011617.2 | - | 18 | 2671 | 3946441 | |
S72370.1 | - | 17 | 2670 | |||
U04641.1 | - | 17 | 2626 | |||
U30891.1 | - | 18 | 2640 |