Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000962.3 | NP_000953.2 | 7 | 772 | ||
NM_001271164.1 | NP_001258093.1 | 6 | 628 | |||
NM_001271165.1 | NP_001258094.1 | 6 | 721 | |||
NM_001271166.1 | NP_001258095.1 | 7 | 998 | |||
NM_001271367.1 | NP_001258296.1 | 7 | 743 | |||
NM_001271368.1 | NP_001258297.1 | 8 | 1220 | |||
NM_080591.2 | NP_542158.1 | 7 | 772 | |||
XM_005252105.3 | XP_005252162.1 | 8 | 947 | |||
XM_011518875.2 | XP_011517177.1 | 8 | 1208 | |||
XM_011518876.2 | XP_011517178.1 | 8 | 4818 | |||
XM_017014929.1 | XP_016870418.1 | 10 | 1182 | |||
GenBank mRNA | AK290022.1 | - | 7 | 725 | ||
AK295221.1 | - | 8 | 1205 | |||
AK304403.1 | - | 6 | 721 | |||
AK308222.1 | - | 7 | 998 | |||
BC029840.1 | - | 7 | 744 | 5174923 | ||
CD014048.1 | - | 6 | 594 | |||
CD014049.1 | - | 7 | 709 | |||
DQ180741.1 | - | 6 | 785 | |||
DQ180742.1 | - | 6 | 785 | |||
M59979.1 | - | 6 | 697 | |||
S36219.1 | - | 6 | 692 | |||
S36271.1 | - | 6 | 692 | |||
S78220.1 | - | 6 | 692 |