Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 4 | 903 | ||
NM_001318368.1 | NP_001305297.1 | 4 | 903 | |||
XM_005266216.4 | XP_005266273.1 | 4 | 261 | |||
XM_011534864.1 | XP_011533166.1 | 4 | 882 | |||
XM_017020317.1 | XP_016875806.1 | 3 | 259 | |||
XM_017020318.1 | XP_016875807.1 | 1 | 51 | |||
GenBank mRNA | AK296875.1 | - | 3 | 246 | ||
AY702976.1 | - | 4 | 365 | |||
BC069634.1 | - | 4 | 388 | 7262421 | ||
BC094784.1 | - | 4 | 903 | |||
BC104492.1 | - | 4 | 391 | 40032854 | ||
BC105950.1 | - | 4 | 391 | 40032852 | ||
BC107601.1 | - | 4 | 390 | 40032849 | ||
X85753.1 | - | 4 | 391 |