Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 6, 7 | 1186 | ||
NM_001318368.1 | NP_001305297.1 | 6, 7 | 1186 | |||
XM_005266216.4 | XP_005266273.1 | 6, 7 | 544 | |||
XM_011534864.1 | XP_011533166.1 | 5, 6 | 1107 | |||
XM_011534865.2 | XP_011533167.1 | 2, 3 | 1231 | |||
XM_017020317.1 | XP_016875806.1 | 5, 6 | 542 | |||
XM_017020318.1 | XP_016875807.1 | 2, 3 | 276 | |||
GenBank mRNA | AK296084.1 | - | 2, 3 | 176 | ||
AK296875.1 | - | 5, 6 | 529 | |||
BC069634.1 | - | 6, 7 | 671 | 7262421 | ||
BC094784.1 | - | 5, 6 | 1128 | |||
BC104492.1 | - | 6, 7 | 674 | 40032854 | ||
BC105950.1 | - | 6, 7 | 674 | 40032852 | ||
BC107601.1 | - | 6, 7 | 673 | 40032849 | ||
X85753.1 | - | 6, 7 | 674 |