Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002615.5 | NP_002606.3 | 4 | 456 | ||
XM_017024754.1 | XP_016880243.1 | 4 | 376 | |||
GenBank mRNA | AB593011.1 | - | 4 | 361 | ||
AB593012.1 | - | 4 | 358 | |||
AB593013.1 | - | 4 | 358 | |||
AF400442.1 | - | 4 | 372 | |||
AK315344.1 | - | 4 | 403 | |||
AY513280.1 | - | 4 | 348 | |||
BC000522.1 | - | 4 | 351 | 2961120 | ||
BC013984.2 | - | 4 | 355 | 3531777 | ||
BT007222.1 | - | 3 | 299 | |||
BU154385.1 | - | 2 | 65 | |||
CA405781.1 | - | 4 | 406 | |||
M90439.1 | - | 2 | 124 |