Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281496.1 | NP_001268425.1 | 7 | 1480 | ||
NM_001281497.1 | NP_001268426.1 | 6 | 1081 | |||
NM_001281715.1 | NP_001268644.1 | 8 | 1524 | |||
NM_002634.3 | NP_002625.1 | 7 | 1432 | |||
XM_017024763.1 | XP_016880252.1 | 8 | 1574 | |||
GenBank mRNA | AK223181.1 | - | 7 | 1407 | ||
BP267961.1 | - | 1 | 315 |