Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002639.4 | NP_002630.2 | 4 | 492 | ||
GenBank mRNA | AK312765.1 | - | 4 | 470 | ||
AL602439.1 | - | 4 | 495 | |||
BC020713.1 | - | 4 | 445 | 4248249 | ||
U04313.1 | - | 4 | 425 |