Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004060.3 | NP_004051.1 | 2, 3 | 506 | ||
NM_199246.1 | NP_954854.1 | 3, 4 | 406 | |||
GenBank mRNA | AK297190.1 | - | 2, 3 | 119 | ||
AK312913.1 | - | 2, 3 | 482 | |||
AK316234.1 | - | 1, 2 | 244 | |||
BC000196.1 | - | 3, 4 | 356 | 3351902 | ||
BC007093.2 | - | 3, 4 | 381 | 4251051 | ||
BM479568.1 | - | 3, 4 | 419 | |||
BT007134.1 | - | 1, 2 | 272 | |||
BX538035.1 | - | 4, 5 | 264 | |||
BX644635.1 | - | 2, 3 | 497 | |||
D78341.1 | - | 2, 3 | 462 | |||
L49504.1 | - | 2, 3 | 483 | |||
U47413.1 | - | 2, 3 | 485 | |||
U53328.1 | - | 1, 2 | 285 | |||
X77794.1 | - | 1, 2 | 254 |