Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 3 | 911 | ||
NM_020473.3 | NP_065206.3 | 2 | 134 | |||
NR_033835.1 | 3 | 537 | ||||
NR_033836.1 | 3 | 253 | ||||
XM_011545539.2 | XP_011543841.1 | 2 | 1735 | |||
XM_017029581.1 | XP_016885070.1 | 2 | 858 | |||
GenBank mRNA | AK098047.1 | - | 2 | 347 | ||
AK290497.1 | - | 2 | 496 | |||
AK303538.1 | - | 2 | 126 | |||
AK304144.1 | - | 3 | 880 | |||
AK315063.1 | - | 2 | 870 | |||
BC038236.1 | - | 2 | 868 | 5733899 | ||
CX870731.1 | - | 2 | 213 | |||
D11466.1 | - | 3 | 880 | |||
DA951065.1 | - | 3 | 253 | |||
DC347872.1 | - | 3 | 537 |
Gene Symbol: PIGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S78467.1 | - | 1 | 860 |