Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018073.2 | NP_001018083.2 | 3 | 609 | ||
NM_001291556.1 | NP_001278485.1 | 3 | 443 | |||
NM_001308054.1 | NP_001294983.1 | 3 | 870 | |||
NM_004563.3 | NP_004554.3 | 3 | 609 | |||
XM_006720158.2 | XP_006720221.1 | 3 | 602 | |||
GenBank mRNA | AK129934.1 | - | 3 | 427 | ||
AK301400.1 | - | 3 | 453 | |||
AK307913.1 | - | 3 | 838 | |||
AK311710.1 | - | 3 | 840 | |||
AK316206.1 | - | 3 | 263 | |||
BC001454.2 | - | 3 | 399 | 3138368 | ||
BX248272.1 | - | 3 | 383 | |||
CR456913.1 | - | 3 | 334 | |||
KU178243.1 | - | 3 | 334 | |||
KU178244.1 | - | 3 | 334 | |||
X92720.1 | - | 3 | 400 |