Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002643.3 | NP_002634.1 | 6 | 843 | ||
NM_173074.2 | NP_775097.1 | 7 | 948 | |||
XM_011532908.2 | XP_011531210.1 | 7 | 830 | |||
GenBank mRNA | AK291108.1 | - | 6 | 798 | ||
BC021725.1 | - | 7 | 888 | 5269689 | ||
BC029408.1 | - | 6 | 796 | 4664848 | ||
D13435.1 | - | 6 | 746 |