Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122752.1 | NP_001116224.1 | 5 | 909 | ||
NM_005025.4 | NP_005016.1 | 5 | 1142 | |||
XM_017006618.1 | XP_016862107.1 | 5 | 1207 | |||
GenBank mRNA | AK290082.1 | - | 5 | 832 | ||
BC018043.1 | - | 5 | 830 | 4796470 | ||
CR627434.1 | - | 5 | 1131 | |||
KU178259.1 | - | 4 | 711 | |||
Z81326.1 | - | 5 | 792 |