Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006835.2 | NP_006826.1 | 7 | 1771 | ||
XM_011531555.1 | XP_011529857.1 | 5 | 1596 | |||
GenBank mRNA | AB209766.1 | - | 7 | 1773 | ||
AK222931.1 | - | 6 | 1223 | |||
BC000420.2 | - | 7 | 1770 | 2964432 | ||
BC004975.1 | - | 7 | 1770 | 2957878 | ||
D50310.1 | - | 6 | 1227 |