Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300960.1 | NP_001287889.1 | 3 | 410 | ||
NM_001300963.1 | NP_001287892.1 | 4 | 226 | |||
NM_001300964.1 | NP_001287893.1 | 3 | 180 | |||
NM_015076.4 | NP_055891.1 | 3 | 410 | |||
XM_011535630.2 | XP_011533932.1 | 4 | 888 | |||
XM_011535632.1 | XP_011533934.1 | 2 | 102 | |||
XM_017010587.1 | XP_016866076.1 | 4 | 636 | |||
XM_017010588.1 | XP_016866077.1 | 2 | 160 | |||
GenBank mRNA | AB028951.2 | - | 3 | 227 | ||
AK292360.1 | - | 3 | 318 | |||
AY028424.1 | - | 3 | 410 | |||
BC024247.1 | - | 4 | 231 | |||
BC037289.1 | - | 3 | 305 | 5266531 | ||
DB030002.1 | - | 3 | 182 | |||
JX028238.1 | - | 3 | 230 |