Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282426.1 | NP_001269355.1 | 2 | 1201 | ||
NM_001282427.1 | NP_001269356.1 | 2 | 1125 | |||
NM_002649.3 | NP_002640.2 | 2 | 1350 | |||
XM_005250443.3 | XP_005250500.1 | 2 | 1218 | |||
XM_011516316.1 | XP_011514618.1 | 2 | 1183 | |||
XM_011516317.1 | XP_011514619.1 | 2 | 1183 | |||
XM_017012328.1 | XP_016867817.1 | 2 | 1183 | |||
GenBank mRNA | AF327656.1 | - | 1 | 1040 | ||
AK292684.1 | - | 2 | 1125 | |||
AK313753.1 | - | 2 | 1108 | |||
BC035683.1 | - | 2 | 1093 | 5749986 | ||
BX648341.1 | - | 2 | 1183 | |||
X83368.1 | - | 2 | 1360 |