Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301668.1 | NP_001288597.1 | 8 | 1344 | ||
NM_015878.5 | NP_056962.2 | 9 | 1490 | |||
NM_148174.3 | NP_680479.1 | 8 | 1344 | |||
XM_005250969.1 | XP_005251026.1 | 8 | 1305 | |||
XM_011517126.1 | XP_011515428.1 | 9 | 1477 | |||
XM_011517127.1 | XP_011515429.1 | 9 | 1468 | |||
XM_011517128.2 | XP_011515430.1 | 8 | 1488 | |||
XM_017013563.1 | XP_016869052.1 | 9 | 1456 | |||
XM_017013564.1 | XP_016869053.1 | 9 | 1468 | |||
XM_017013565.1 | XP_016869054.1 | 9 | 1607 | |||
XM_017013566.1 | XP_016869055.1 | 8 | 1488 | |||
GenBank mRNA | AK095493.1 | - | 8 | 1272 | ||
AK290516.1 | - | 9 | 1222 | |||
BC013420.2 | - | 9 | 1429 | 3509827 | ||
BC019279.1 | - | 8 | 1291 | 2823000 | ||
BC042404.1 | - | 8 | 1315 | |||
BT006858.1 | - | 6 | 717 | |||
CR456745.1 | - | 6 | 717 | |||
D88674.1 | - | 9 | 1426 |