Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113575.1 | NP_001107047.1 | 4 | 629 | ||
NM_001300853.1 | NP_001287782.1 | 3 | 395 | |||
NM_016508.3 | NP_057592.2 | 4 | 629 | |||
XM_011543429.2 | XP_011541731.1 | 4 | 944 | |||
XM_017009524.1 | XP_016865013.1 | 3 | 1376 | |||
XM_017009525.1 | XP_016865014.1 | 4 | 944 | |||
XM_017009526.1 | XP_016865015.1 | 4 | 942 | |||
XM_017009527.1 | XP_016865016.1 | 3 | 1376 | |||
XM_017009528.1 | XP_016865017.1 | 3 | 1376 | |||
XM_017009529.1 | XP_016865018.1 | 3 | 1376 | |||
XM_017009530.1 | XP_016865019.1 | 3 | 1376 | |||
XM_017009531.1 | XP_016865020.1 | 3 | 1376 | |||
XM_017009532.1 | XP_016865021.1 | 3 | 1375 | |||
XM_017009533.1 | XP_016865022.1 | 3 | 1375 | |||
XM_017009534.1 | XP_016865023.1 | 3 | 1375 | |||
XM_017009535.1 | XP_016865024.1 | 3 | 1375 | |||
XM_017009536.1 | XP_016865025.1 | 3 | 1377 | |||
XM_017009538.1 | XP_016865027.1 | 3 | 420 | |||
XM_017009544.1 | XP_016865033.1 | 3 | 420 | |||
XM_017009545.1 | XP_016865034.1 | 2 | 227 | |||
XM_017009546.1 | XP_016865035.1 | 2 | 220 | |||
XM_017009548.1 | XP_016865037.1 | 3 | 1380 | |||
GenBank mRNA | AF130372.1 | - | 3 | 510 | ||
AK301802.1 | - | 3 | 389 | |||
BC041799.1 | - | 4 | 631 | 5298279 |