Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184779.1 | NP_001171708.1 | 9 | 1382 | ||
NM_001184780.1 | NP_001171709.1 | 9 | 1509 | |||
NM_024505.3 | NP_078781.3 | 9 | 1466 | |||
NR_033671.2 | 10 | 1697 | ||||
NR_033672.1 | 10 | 1613 | ||||
GenBank mRNA | AF317889.1 | - | 6 | 897 | ||
AF325189.1 | - | 10 | 1588 | |||
AF325190.1 | - | 10 | 1672 | |||
AF353088.1 | - | 9 | 1361 | |||
AF353089.1 | - | 9 | 1445 | |||
AK026011.1 | - | 6 | 951 | |||
AK074058.1 | - | 7 | 1135 | |||
AK074071.1 | - | 8 | 4584 | |||
AK314689.1 | - | 9 | 1445 | |||
BC125097.1 | - | 9 | 1375 | 40118286 | ||
BC125098.1 | - | 10 | 1479 | 40118288 | ||
DQ413001.1 | - | 6 | 825 |