Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253875.1 | NP_001240804.1 | 15 | 1280 | ||
NM_001253876.1 | NP_001240805.1 | 10 | 1417 | |||
NM_025076.4 | NP_079352.2 | 15 | 1265 | |||
NR_045607.1 | 14 | 1216 | ||||
XM_006712775.3 | XP_006712838.1 | 14 | 1138 | |||
XM_011511902.2 | XP_011510204.2 | 15 | 1259 | |||
XM_011511903.2 | XP_011510205.2 | 15 | 1242 | |||
XM_011511904.2 | XP_011510206.1 | 12 | 1587 | |||
XM_017005008.1 | XP_016860497.1 | 13 | 1080 | |||
XM_017005009.1 | XP_016860498.1 | 13 | 1065 | |||
XM_017005010.1 | XP_016860499.1 | 14 | 1123 | |||
XM_017005011.1 | XP_016860500.1 | 15 | 1367 | |||
XM_017005012.1 | XP_016860501.1 | 15 | 1302 | |||
XM_017005013.1 | XP_016860502.1 | 13 | 1825 | |||
XM_017005014.1 | XP_016860503.1 | 14 | 1043 | |||
GenBank mRNA | AK027244.1 | - | 10 | 1417 | ||
AK056022.1 | - | 4 | 2094 | |||
AK075120.1 | - | 15 | 1227 | |||
AK075170.1 | - | 15 | 1242 | |||
AK122696.1 | - | 14 | 1181 | |||
AK225592.1 | - | 10 | 1417 | |||
AK290668.1 | - | 15 | 1240 | |||
AL834439.1 | - | 2 | 1029 | |||
AY147934.1 | - | 15 | 1232 | |||
AY358541.1 | - | 15 | 1216 | |||
BC009819.2 | - | 15 | 1206 | 3843312 |