Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030666.3 | NP_109591.1 | 5 | 789 | ||
NR_073111.1 | 6 | 921 | ||||
NR_073112.1 | 5 | 789 | ||||
XM_011514333.1 | XP_011512635.1 | 6 | 1012 | |||
XM_011514334.2 | XP_011512636.1 | 5 | 923 | |||
XM_011514335.2 | XP_011512637.1 | 5 | 778 | |||
GenBank mRNA | AK223370.1 | - | 5 | 602 | ||
AK291327.1 | - | 5 | 602 | |||
AK298044.1 | - | 6 | 731 | |||
AK304641.1 | - | 5 | 634 | |||
AK312973.1 | - | 5 | 600 | |||
BC009015.1 | - | 5 | 587 | 3456154 | ||
BI521531.1 | - | 3 | 260 | |||
BT006928.1 | - | 4 | 543 | |||
BU942473.1 | - | 5 | 604 | |||
EU794624.1 | - | 4 | 543 | |||
GQ472232.1 | - | 5 | 587 | |||
M93056.1 | - | 5 | 591 |