Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053056.2 | NP_444284.1 | 5 | 2452 | ||
GenBank mRNA | AB075505.1 | - | 1 | 41 | ||
BC023620.2 | - | 5 | 2379 | 4650919 | ||
M73554.1 | - | 5 | 2384 | |||
X59798.1 | - | 5 | 2390 |