Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324081.1 | NP_001311010.1 | 7 | 1428 | ||
NM_080861.3 | NP_543137.2 | 7 | 1450 | |||
XM_005255673.4 | XP_005255730.1 | 7 | 1640 | |||
GenBank mRNA | AK074190.1 | - | 7 | 1414 | ||
BC007588.1 | - | 7 | 1396 | 3342825 | ||
BC024244.1 | - | 7 | 1414 | 4798873 | ||
BC035065.1 | - | 7 | 1407 | |||
BC041897.1 | - | 6 | 2224 | 5298729 | ||
BC047441.1 | - | 7 | 1392 | 5303706 | ||
BC065515.1 | - | 4 | 1693 |