Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198834.2 | NP_942131.1 | 18 | 2793 | ||
NM_198836.2 | NP_942133.1 | 18 | 2361 | |||
NM_198837.1 | NP_942134.1 | 16 | 2431 | |||
NM_198838.1 | NP_942135.1 | 17 | 2542 | |||
NM_198839.2 | NP_942136.1 | 22 | 3467 | |||
XM_005257267.4 | XP_005257324.1 | 18 | 2274 | |||
XM_006721853.1 | XP_006721916.1 | 18 | 2804 | |||
XM_011524701.1 | XP_011523003.1 | 18 | 2714 | |||
XM_011524703.1 | XP_011523005.1 | 18 | 2346 | |||
XM_011524704.2 | XP_011523006.1 | 18 | 2279 | |||
XM_017024553.1 | XP_016880042.1 | 19 | 2828 | |||
XM_017024554.1 | XP_016880043.1 | 19 | 2828 | |||
XM_017024555.1 | XP_016880044.1 | 18 | 2427 | |||
GenBank mRNA | AB371587.1 | - | 18 | 2340 | ||
AK309084.1 | - | 18 | 2349 | |||
AY237919.1 | - | 16 | 2170 | |||
AY315627.1 | - | 16 | 2170 | |||
BC137287.1 | - | 18 | 2298 | 9021284 |