Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001215.3 | NP_001206.2 | 6 | 691 | ||
NM_001270500.1 | NP_001257429.1 | 6 | 691 | |||
NM_001270501.1 | NP_001257430.1 | 5 | 511 | |||
NM_001270502.1 | NP_001257431.1 | 4 | 362 | |||
XM_011542083.2 | XP_011540385.1 | 6 | 686 | |||
XM_011542084.2 | XP_011540386.1 | 6 | 739 | |||
GenBank mRNA | AB102863.1 | - | 6 | 638 | ||
AB103091.1 | - | 5 | 458 | |||
AK300652.1 | - | 6 | 655 | |||
CB956732.1 | - | 4 | 331 | |||
M57892.1 | - | 6 | 644 |