Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001215.3 | NP_001206.2 | 8 | 1250 | ||
NM_001270501.1 | NP_001257430.1 | 7 | 1070 | |||
NM_001270502.1 | NP_001257431.1 | 6 | 921 | |||
GenBank mRNA | BX282262.1 | - | 1 | 194 | ||
M57892.1 | - | 8 | 1203 |