Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001218.4 | NP_001209.1 | 8 | 1246 | ||
NM_001293642.1 | NP_001280571.1 | 7 | 1066 | |||
NM_206925.2 | NP_996808.1 | 8 | 1246 | |||
NR_135511.1 | 8 | 1349 | ||||
GenBank mRNA | AF037335.1 | - | 8 | 971 | ||
AF051882.1 | - | 8 | 863 | |||
AK096845.1 | - | 7 | 802 | |||
AK315769.1 | - | 8 | 1012 | |||
BC000278.1 | - | 8 | 968 | 3357439 | ||
BC011691.2 | - | 8 | 971 | 3344734 | ||
BC023981.1 | - | 8 | 986 | 4042589 | ||
BT006656.1 | - | 8 | 856 |