Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321839.1 | NP_001308768.1 | 7, 8 | 1099 | ||
NM_004056.5 | NP_004047.3 | 8, 9 | 1195 | |||
NR_135821.1 | 9, 10 | 1245 | ||||
XM_017013818.1 | XP_016869307.1 | 9, 10 | 1260 | |||
GenBank mRNA | AK090655.1 | - | 8, 9 | 1167 | ||
AK293999.1 | - | 7, 8 | 786 | |||
BC069744.1 | - | 8, 9 | 1164 | 7262540 | ||
BC069794.1 | - | 8, 9 | 1199 | 7262552 | ||
BC108929.2 | - | 8, 9 | 1120 | 40025442 |