Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005181.3 | NP_005172.1 | 4 | 463 | ||
GenBank mRNA | AK096880.1 | - | 4 | 463 | ||
AK300670.1 | - | 4 | 357 | |||
AK300677.1 | - | 3 | 384 | |||
AK313254.1 | - | 4 | 432 | |||
BC004897.2 | - | 4 | 418 | 3532715 | ||
DA421884.1 | - | 4 | 463 | |||
EU794596.1 | - | 4 | 1063 |