Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130082.2 | NP_001123554.1 | 3 | 825 | ||
NM_002673.5 | NP_002664.2 | 3 | 992 | |||
XM_011533833.2 | XP_011532135.1 | 3 | 878 | |||
XM_011533834.1 | XP_011532136.1 | 3 | 867 | |||
XM_011533835.1 | XP_011532137.1 | 3 | 884 | |||
XM_011533836.1 | XP_011532138.1 | 3 | 800 | |||
XM_011533837.2 | XP_011532139.1 | 3 | 973 | |||
XM_017006630.1 | XP_016862119.1 | 3 | 800 | |||
XM_017006631.1 | XP_016862120.1 | 2 | 4410 | |||
GenBank mRNA | AB007867.1 | - | 3 | 992 | ||
AJ011414.1 | - | 1 | 722 | |||
AJ011415.1 | - | 1 | 722 | |||
BC146793.1 | - | 3 | 993 | 9007264 | ||
X87904.2 | - | 1 | 722 |