Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042.2 | NP_001033.1 | 11 | 1667 | ||
GenBank mRNA | AK313372.1 | - | 11 | 1681 | ||
BC034387.1 | - | 11 | 1728 | 5187454 | ||
BC069615.1 | - | 11 | 1562 | 7262457 | ||
BC069621.1 | - | 11 | 1562 | 7262469 | ||
BC113592.1 | - | 11 | 1494 | 8322644 | ||
BC126164.1 | - | 11 | 1494 | 8991880 | ||
M20747.1 | - | 11 | 1612 |