Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003039.2 | NP_003030.1 | 12 | 2026 | ||
XM_005263491.3 | XP_005263548.1 | 13 | 1824 | |||
XM_017002133.1 | XP_016857622.1 | 14 | 2052 | |||
XM_017002134.1 | XP_016857623.1 | 14 | 2018 | |||
XM_017002135.1 | XP_016857624.1 | 14 | 2112 | |||
XM_017002136.1 | XP_016857625.1 | 14 | 2088 | |||
XM_017002137.1 | XP_016857626.1 | 15 | 2124 | |||
XM_017002138.1 | XP_016857627.1 | 15 | 2100 | |||
XM_017002139.1 | XP_016857628.1 | 13 | 2034 | |||
XM_017002140.1 | XP_016857629.1 | 14 | 1955 | |||
XM_017002141.1 | XP_016857630.1 | 13 | 1857 | |||
XM_017002142.1 | XP_016857631.1 | 10 | 1597 | |||
GenBank mRNA | AK294368.1 | - | 12 | 1812 | ||
AK300477.1 | - | 10 | 1602 | |||
AK302057.1 | - | 15 | 1950 | |||
AK302137.1 | - | 14 | 1902 | |||
BC001692.2 | - | 12 | 1785 | 2966459 | ||
BC001820.2 | - | 12 | 1785 | 2966459 | ||
M55531.1 | - | 12 | 1809 |