Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018711.4 | NP_061181.1 | 9 | 1219 | ||
GenBank mRNA | BC033587.1 | - | 9 | 1019 | 5243017 | |
BC094722.1 | - | 9 | 866 | 4817829 |