Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001290.1 | NP_001001290.1 | 7 | 875 | ||
NM_020041.2 | NP_064425.2 | 6 | 864 | |||
XM_006713968.3 | XP_006714031.1 | 6 | 976 | |||
XM_006713969.2 | XP_006714032.1 | 7 | 837 | |||
XM_011513856.2 | XP_011512158.1 | 6 | 1001 | |||
XM_011513857.1 | XP_011512159.1 | 7 | 837 | |||
XM_011513858.1 | XP_011512160.1 | 7 | 1039 | |||
XM_011513859.2 | XP_011512161.1 | 6 | 971 | |||
XM_011513860.2 | XP_011512162.1 | 6 | 971 | |||
XM_011513861.2 | XP_011512163.1 | 6 | 973 | |||
XM_011513862.2 | XP_011512164.1 | 5 | 848 | |||
XM_011513863.2 | XP_011512165.1 | 5 | 2527 | |||
XM_011513864.2 | XP_011512166.1 | 4 | 416 | |||
XM_011513865.2 | XP_011512167.1 | 6 | 975 | |||
XM_011513866.2 | XP_011512168.1 | 6 | 973 | |||
XM_011513867.2 | XP_011512169.1 | 3 | 930 | |||
XM_011513868.2 | XP_011512170.1 | 6 | 992 | |||
XM_017008457.1 | XP_016863946.1 | 6 | 989 | |||
XM_017008458.1 | XP_016863947.1 | 6 | 974 | |||
XM_017008459.1 | XP_016863948.1 | 4 | 508 | |||
XM_017008460.1 | XP_016863949.1 | 5 | 2521 | |||
GenBank mRNA | AF210317.1 | - | 6 | 857 | ||
AF421859.1 | - | 4 | 552 | |||
AK310895.1 | - | 7 | 837 | |||
BC018897.2 | - | 7 | 875 | 3949549 | ||
BC110414.1 | - | 6 | 894 | 5762360 |