Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003486.5 | NP_003477.4 | 10 | 2013 | ||
XM_006721286.3 | XP_006721349.2 | 10 | 1656 | |||
GenBank mRNA | AB018009.1 | - | 10 | 2006 | ||
AF104032.1 | - | 10 | 2007 | |||
BC039692.2 | - | 10 | 2036 | 5551612 | ||
BC042600.1 | - | 10 | 2007 | 4470593 | ||
M80244.1 | - | 10 | 1452 |