Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001998.2 | NP_001001998.1 | 2, 3 | 323 | ||
NM_002685.3 | NP_002676.1 | 2, 3 | 323 | |||
XM_005263475.2 | XP_005263532.2 | 2, 3 | 282 | |||
XM_017001494.1 | XP_016856983.1 | 2, 3 | 199 | |||
XM_017001495.1 | XP_016856984.1 | 2, 3 | 199 | |||
XM_017001496.1 | XP_016856985.1 | 2, 3 | 199 | |||
XM_017001497.1 | XP_016856986.1 | 2, 3 | 199 | |||
GenBank mRNA | AB209236.1 | - | 2, 3 | 242 | ||
AK294058.1 | - | 2, 3 | 250 | |||
AK296559.1 | - | 2, 3 | 250 | |||
AK308778.1 | - | 2, 3 | 259 | |||
BC039901.1 | - | 2, 3 | 264 | 5505500 | ||
BC073788.1 | - | 2, 3 | 299 | 5420763 | ||
DA505395.1 | - | 2, 3 | 323 | |||
L01457.1 | - | 2, 3 | 282 | |||
X66113.1 | - | 2, 3 | 281 |