Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000814.5 | NP_000805.1 | 8 | 1166 | ||
NM_001191320.1 | NP_001178249.1 | 6 | 964 | |||
NM_001191321.2 | NP_001178250.1 | 6 | 1014 | |||
NM_001278631.1 | NP_001265560.1 | 9 | 1262 | |||
NM_021912.4 | NP_068712.1 | 8 | 1116 | |||
XM_011521428.2 | XP_011519730.1 | 7 | 1149 | |||
GenBank mRNA | AK295167.1 | - | 6 | 930 | ||
AK302822.1 | - | 6 | 964 | |||
AK315311.1 | - | 8 | 1117 | |||
BC010641.1 | - | 8 | 1132 | 3871111 | ||
CD013894.1 | - | 9 | 1201 | |||
KJ534842.1 | - | 8 | 1052 | |||
M82919.1 | - | 8 | 1105 |