Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271998.1 | NP_001258927.1 | 3 | 399 | ||
NM_004578.3 | NP_004569.2 | 5 | 633 | |||
NR_073545.1 | 5 | 595 | ||||
GenBank mRNA | AF498934.1 | - | 5 | 355 | ||
AK223009.1 | - | 5 | 398 | |||
AK313807.1 | - | 5 | 606 | |||
AY585832.1 | - | 5 | 562 | |||
BC002438.1 | - | 5 | 426 | 3346455 | ||
BC004309.1 | - | 5 | 563 | 3627110 | ||
BI828974.1 | - | 5 | 479 | |||
BX354553.2 | - | 3 | 328 | |||
KC407675.1 | - | 5 | 317 | |||
M28211.1 | - | 5 | 410 | |||
X13902.1 | - | 5 | 466 |