Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000665.4 | NP_000656.1 | 4 | 1815 | ||
NM_001282449.1 | NP_001269378.1 | 4 | 1551 | |||
NM_001302621.1 | NP_001289550.1 | 4 | 1818 | |||
NM_001302622.1 | NP_001289551.1 | 4 | 1720 | |||
NM_015831.2 | NP_056646.1 | 4 | 1764 | |||
XM_006715995.2 | XP_006716058.1 | 4 | 1720 | |||
XM_011516225.1 | XP_011514527.1 | 3 | 2171 | |||
XM_011516226.1 | XP_011514528.1 | 3 | 1907 | |||
XM_011516228.2 | XP_011514530.1 | 4 | 1723 | |||
XM_011516229.2 | XP_011514531.1 | 4 | 1664 | |||
XM_017012219.1 | XP_016867708.1 | 3 | 2171 | |||
XM_017012220.1 | XP_016867709.1 | 3 | 1907 | |||
GenBank mRNA | AF334270.1 | - | 4 | 1466 | ||
AK223443.1 | - | 4 | 1729 | |||
AK291321.1 | - | 4 | 1715 | |||
BC026315.1 | - | 4 | 1732 | |||
BC036813.1 | - | 4 | 1733 | |||
BC105060.1 | - | 4 | 1708 | 8144063 | ||
BC105062.1 | - | 4 | 1708 | 8144065 | ||
BC143469.1 | - | 4 | 1708 | 9051977 | ||
M55040.1 | - | 3 | 1781 |