Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144829.2 | NP_001138301.1 | 7 | 1322 | ||
NM_001461.3 | NP_001452.2 | 7 | 1312 | |||
XM_005272946.4 | XP_005273003.1 | 7 | 1286 | |||
XM_005272947.4 | XP_005273004.1 | 7 | 1030 | |||
XM_005272948.4 | XP_005273005.1 | 7 | 1013 | |||
XM_011509350.2 | XP_011507652.1 | 7 | 1317 | |||
XM_011509351.2 | XP_011507653.1 | 6 | 1049 | |||
XM_017000801.1 | XP_016856290.1 | 6 | 1061 | |||
XM_017000802.1 | XP_016856291.1 | 6 | 831 | |||
GenBank mRNA | AK222728.1 | - | 7 | 1027 | ||
AK225739.1 | - | 7 | 1037 | |||
AK314647.1 | - | 7 | 1240 | |||
L37080.1 | - | 7 | 998 | |||
Z47553.1 | - | 7 | 1004 |