Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000129.3 | NP_000120.2 | 4 | 673 | ||
GenBank mRNA | AB208852.1 | - | 4 | 625 | ||
AK304335.1 | - | 3 | 455 | |||
AK312735.1 | - | 4 | 644 | |||
BC027963.1 | - | 4 | 631 | 5222962 | ||
M14354.1 | - | 4 | 634 | |||
M14539.1 | - | 4 | 637 |