Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000147.4 | NP_000138.2 | 3, 4 | 753 | ||
XM_005245821.2 | XP_005245878.1 | 3, 4 | 890 | |||
XM_011541167.2 | XP_011539469.1 | 3, 4 | 338 | |||
XM_017000905.1 | XP_016856394.1 | 3, 4 | 435 | |||
GenBank mRNA | AB074175.1 | - | 3, 4 | 674 | ||
AK092914.1 | - | 4, 5 | 608 | |||
AK095443.1 | - | 4, 5 | 625 | |||
AK095455.1 | - | 4, 5 | 639 | |||
AK225468.1 | - | 3, 4 | 674 | |||
AK295535.1 | - | 3, 4 | 857 | |||
AK301302.1 | - | 5, 6 | 603 | |||
AK310143.1 | - | 4, 5 | 736 | |||
AK314649.1 | - | 3, 4 | 674 | |||
BC017338.2 | - | 3, 4 | 676 | 4871788 | ||
M10355.1 | - | 3, 4 | 448 | |||
M29877.1 | - | 3, 4 | 673 | |||
X01390.1 | - | 3, 4 | 448 |