Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 6 | 1177 | ||
NM_001284290.1 | NP_001271219.1 | 4 | 739 | |||
NM_001293104.1 | NP_001280033.1 | 5 | 992 | |||
NM_001293105.1 | NP_001280034.1 | 4 | 849 | |||
NR_120531.1 | 6 | 1177 | ||||
XM_011516113.1 | XP_011514415.1 | 5 | 921 | |||
XM_011516114.1 | XP_011514416.1 | 5 | 849 | |||
GenBank mRNA | AK096764.1 | - | 1 | 1222 | ||
AK223406.1 | - | 6 | 1076 | |||
AK303819.1 | - | 4 | 636 | |||
AK312752.1 | - | 6 | 1093 | |||
BC014142.2 | - | 6 | 1047 | 4662011 | ||
CD014092.1 | - | 5 | 874 | |||
CD014093.1 | - | 4 | 732 | |||
CD014094.1 | - | 6 | 1059 | |||
M15182.1 | - | 6 | 1072 |