Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000262.2 | NP_000253.1 | 7, 8 | 1500 | ||
XM_005261615.4 | XP_005261672.1 | 8, 9 | 1139 | |||
XM_005261616.4 | XP_005261673.1 | 8, 9 | 1121 | |||
GenBank mRNA | BC000095.2 | - | 7, 8 | 1036 | 3504221 | |
CR456527.1 | - | 7, 8 | 1009 | |||
M29276.1 | - | 8 | 1368 | |||
M38083.1 | - | 7, 8 | 1027 | |||
M62783.1 | - | 7, 8 | 1424 |