Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000305.2 | NP_000296.2 | 9 | 1283 | ||
NM_001018161.1 | NP_001018171.1 | 9 | 1247 | |||
XM_005250453.1 | XP_005250510.1 | 9 | 1213 | |||
XM_017012357.1 | XP_016867846.1 | 8 | 1132 | |||
XM_017012358.1 | XP_016867847.1 | 8 | 1158 | |||
GenBank mRNA | AF001601.1 | - | 9 | 1195 | ||
AK054688.1 | - | 1 | 1829 | |||
AK291103.1 | - | 9 | 1205 | |||
AK296029.1 | - | 9 | 1162 | |||
AK296636.1 | - | 9 | 1203 | |||
AK301313.1 | - | 9 | 1410 | |||
AK308065.1 | - | 9 | 1214 | |||
BC040010.1 | - | 9 | 1186 | 6056277 | ||
BC046160.1 | - | 9 | 1149 | 5393700 | ||
L48513.1 | - | 9 | 1176 |
Gene Symbol: ZFHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR933619.1 | - | 5534 |