Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 6 | 820 | ||
NM_001079811.2 | NP_001073279.1 | 6 | 714 | |||
NM_001317040.1 | NP_001303969.1 | 7 | 964 | |||
GenBank mRNA | AK130133.1 | - | 6 | 748 | ||
AK222763.1 | - | 6 | 711 | |||
AK300021.1 | - | 6 | 647 | |||
AK300753.1 | - | 7 | 880 | |||
AK312988.1 | - | 6 | 707 | |||
BC007493.2 | - | 6 | 722 | 2988086 | ||
BT007147.1 | - | 6 | 675 | |||
M22590.1 | - | 6 | 709 | |||
M27507.1 | - | 6 | 725 | |||
M34423.1 | - | 6 | 735 |