Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000512.4 | NP_000503.1 | 4 | 501 | ||
NM_001323543.1 | NP_001310472.1 | 3 | 381 | |||
NM_001323544.1 | NP_001310473.1 | 5 | 675 | |||
XM_005256301.3 | XP_005256358.1 | 4 | 501 | |||
XM_011522982.2 | XP_011521284.1 | 5 | 547 | |||
XM_017023111.1 | XP_016878600.1 | 4 | 2398 | |||
XM_017023112.1 | XP_016878601.1 | 4 | 2398 | |||
XM_017023113.1 | XP_016878602.1 | 3 | 359 | |||
GenBank mRNA | AK131096.1 | - | 2 | 3820 | ||
AK294390.1 | - | 2 | 251 | |||
AK308269.1 | - | 5 | 531 | |||
AK310392.1 | - | 5 | 547 | |||
AK312655.1 | - | 4 | 478 | |||
AY129021.1 | - | 2 | 178 | |||
BC050684.1 | - | 4 | 490 | 6091623 | ||
BC056151.1 | - | 4 | 482 | 6026248 | ||
BX649181.1 | - | 3 | 238 | |||
DA010361.1 | - | 4 | 501 | |||
DA149503.1 | - | 3 | 358 | |||
DA709138.1 | - | 4 | 505 |