Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000792.6 | NP_000783.2 | 3 | 676 | ||
NM_001039715.2 | NP_001034804.1 | 2 | 532 | |||
NM_213593.4 | NP_998758.1 | 3 | 484 | |||
NR_136692.1 | 3 | 567 | ||||
NR_136693.1 | 4 | 593 | ||||
GenBank mRNA | AK290780.1 | - | 3 | 676 | ||
AY560374.1 | - | 3 | 652 | |||
AY560375.1 | - | 3 | 460 | |||
AY560377.1 | - | 2 | 508 | |||
AY560379.1 | - | 3 | 543 | |||
AY560380.1 | - | 4 | 528 | |||
AY560382.1 | - | 4 | 569 | |||
AY560383.1 | - | 3 | 425 | |||
BC107170.2 | - | 3 | 675 | 40036824 | ||
BC107171.2 | - | 3 | 675 | 40036826 | ||
FJ002243.1 | - | 4 | 591 | |||
FJ002244.1 | - | 3 | 565 | |||
FJ002249.1 | - | 3 | 64 |