Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000793.5 | NP_000784.2 | 4 | 703 | ||
NM_001007023.4 | NP_001007024.1 | 3 | 580 | |||
NM_001242502.2 | NP_001229431.1 | 4 | 714 | |||
NM_001242503.2 | NP_001229432.1 | 3 | 606 | |||
NM_001324462.1 | NP_001311391.1 | 3 | 719 | |||
NM_013989.4 | NP_054644.1 | 2 | 472 | |||
GenBank mRNA | AB041843.1 | - | 3 | 453 | ||
AB041844.1 | - | 4 | 400 | |||
AF093774.1 | - | 2 | 472 | |||
AK292210.1 | - | 4 | 703 | |||
AK311603.1 | - | 3 | 293 | |||
BC063118.1 | - | 2 | 355 | |||
BC074882.2 | - | 2 | 348 | 30915240 | ||
BC136514.1 | - | 2 | 342 | 9020504 | ||
CD691767.1 | - | 2 | 406 | |||
JK546617.1 | - | 3 | 630 | |||
U53506.1 | - | 2 | 451 |