Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146258.1 | NP_001139730.1 | 2 | 370 | ||
NM_001146259.1 | NP_001139731.1 | 2 | 370 | |||
NM_001146260.1 | NP_001139732.1 | 2 | 370 | |||
NM_024779.4 | NP_079055.3 | 2 | 370 | |||
XM_005269152.2 | XP_005269209.1 | 2 | 363 | |||
XM_011538747.1 | XP_011537049.1 | 2 | 363 | |||
XM_017019973.1 | XP_016875462.1 | 2 | 327 | |||
GenBank mRNA | AK075553.1 | - | 2 | 326 | ||
AK125526.1 | - | 2 | 254 | |||
AK297243.1 | - | 2 | 355 | |||
AK302254.1 | - | 2 | 286 | |||
AK315588.1 | - | 2 | 250 | |||
AX754970.1 | - | 2 | 311 | |||
BC028596.2 | - | 2 | 255 | 4824916 | ||
BX419431.2 | - | 2 | 368 |